À propos de cet article
MPure Tissue DNA Extraction Kit is used with the MPure-12 nucleic acid purification system for the extraction and purification of genomic DNA from a variety of animal tissues, swab samples and blood stain.
- Extraction of genomic DNA from animal tissues, swab samples, and blood stains
- Completely automated purification protocol
- Use with MPure-12 System
- Run in as little as 40 minutes
- Nucleic Acids can be used immediately in downstream applications
MPure™ Tissue DNA Extraction Kit contains everything needed for sample preparation. However, for tough tissues such as cardiac or skeletal muscle, spleen, intestinal or other connective tissue-rich samples, sample preparation using FastPrep™ Instruments and Lysing Matrix Tubes can greatly increase yields as well as shorten processing time.
Capacity: 100-400 µL / 1 swab or brush for Dried Swab samples, 100-400 µL / 4 discs for dried blood, Purify DNA from 100-400 µL / 10-40mg of tissue
Sample Type: Animal Tissues, Blood Stain, Swab
Sample Volume: 100-400 µL
Spécifications
- Method / Format:Magnetic beads
- Prep size:Mini
- Refine target molecule:Total DNA
- Sample type:Cultured cellsHuman/animal cells and tissues
- Target molecule:DNA
- Description:MPure™ Tissue DNA Extraction Kit