MPurify Your Nucleic Acid: Obtain Superior Yield, Exceptional Purity, and Automated Performance in a Flexible, Convenient and Affordable Manner!
- Extraction of host gDNA, mitochondrial and viral DNA
- Extract DNA from 100-400µL mammalian whole blood
- Nucleic acids can be used immediately in downstream applications
- Use with MPure-12 System
- Completely automated
- Can be run un as little as 50 minutes
MPure Blood DNA Extraction Kit 200 is used with the MPure-12 aNAP System for extraction of DNA from 100-400 µL mammalian whole blood, suspension of mammalian blood cells. Nucleic acids extracted from MPure Blood DNA Extraction Kit 200 can be used in a number of downstream application including: PCR, qPCR, Sequencing (NGS), Microarray, RFLP, Southern Blot Analysis.
Content : Proteinase K solution = 40 µL, Lysis Buffer 2 = 1000 µL, Binding Buffer 1 = 600 µL, Magnetic Bead Solution = 800 µL, Washing Buffer 1 = 1000 µL, Washing Buffer 2 = 1000 µL, Washing Buffer 3 = 1000 µL, Elution Buffer 1 = 1000 µL, Elution Buffer 2 = 1000 µL
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Method / Format:Magnetic beads
- Prep size:Mini
- Refine target molecule:Total DNA
- Sample type:Blood
- Target molecule:DNA
- Sample size:100-400 µL whole blood (WBC count less than 2x 10000 cells/µL) ; 100-400µL leukocyte concentration (contain no more than 5 x 10^6 cells) ; 100-400µL buffy coat. As little of 5 IU spiked (about 1 IU in PCR reaction) sample can be detected.
- Description:MPure™ Blood DNA Extraction Kit 200