Open-Head UN Rated Poly Drum, PIG®, New Pig
Catalog # 89370-998
Supplier: New Pig
Specifications
- Capacity:55 gal.
- Description:Open-Head UN Rated Poly Drum
- Cat. no.:89370-998
- Supplier No.:DRM1147
Specifications
About this item
Ship chemicals in these sturdy, lightweight and rustproof HDPE drums.
- Rustproof
- Rigid, high-density polyethylene is lightweight
- Compatible with a wide range of liquids and chemicals
Ship chemicals in these sturdy, lightweight and rustproof HDPE drums - you can even move them with a drum lifter! Rigid, high-density polyethylene is lightweight and won't rust. Compatible with a wide range of materials, liquids and chemicals.