STAT-MAT® Absorbent, PIG®
PIG® STAT-MAT® Absorbent, 40.2 gal./roll
Catalog # 56620-542
Supplier: New Pig
Supplier #: MAT212
Jump to:
Product Details & Documents
Heavyweight for increased durability and absorbency.
- Dissipates static buildup
- Deters static charge during transportation
- Increased durability and absorbency
Heavyweight for increased durability and absorbency. Treated with a topical solution to dissipate static buildup. Packaged in special static-dissipative bags to deter a static charge during transportation.
Specifications
- Désignation:PIG® STAT-MAT® Absorbent
- Capacité d'absorption:40.2 gal./roll