Absorb-&-Lock® Bio-Fluids Absorbent, PIG®
PIG® Absorb-&-Lock® Bio-Fluids Absorbent, 4 containers per box, 8 gal./container
Catalog # 10861-014
Supplier: New Pig
Fournisseur #: PLP503
Jump to:
Product Details & Documents
Permanently solidifies to lock in absorbed liquids so they won't leak or liquify when the absorbents are handled or placed under pressure.
- Contains chlorine to kill pathogens
- Clean scent helps reduce offensive odors
- Environmentally-friendly product
Contains chlorine to kill pathogens, reduce odors and help you meet OSHA standards. Clean scent helps reduce offensive odors during and after cleanup. Environmentally-friendly product absorbs biological fluids including blood, urine and vomit.
Spécifications
- Désignation:PIG® Absorb-&-Lock® Bio-Fluids Absorbent
- Taille:4 containers per box
- Capacité d'absorption:8 gal./container
- Cat. No.:10861-014