Antimony Standard for AAS, Certipur®, MilliporeSigma
Supplier: MilliporeSigma
Certificates
About this item
Sb₂O₃ in HCl
Traceable to SRM from NIST.
Specifications
- Hazard class description:Corrosive Materials