Retract-a-Clamp® Grounding Assembly, PIG®
Catalog # 10860-606
Supplier: New Pig
Specifications
- Description:Retract-a-Clamp® Grounding Assembly
- Cat. no.:10860-606
- Supplier No.:DRM441
Specifications
About this item
Bonding and grounding assembly controls static electricity to prevent sparking while transferring or storing flammable liquids.
- Solid grounding connection while staying coiled
- Controls static electricity
- Vinyl coating protects wire from moisture
Wire creates a solid grounding connection while staying coiled so as not to be a tripping hazard. Vinyl coating protects wire from moisture and corrosion for longer life and to allow outdoor use.
Certifications: FM approved and NFPA 77 compliant.