ContactUsLinkComponent
Leak Diverter for Spraying Pipes, PIG®
  10860-096
 :  New Pig
undefined
Leak Diverter for Spraying Pipes, PIG®
  10860-096
 :  New Pig
 :  TLS147-YW
  • Color:
    Yellow
  • Description:
    PIG® Leak Diverter for Spraying Pipes
  • Dimensions:
    18" W x 30" L
  • Cat. no.:
    10860-096
  • Supplier No.:
    TLS147-YW
Material wraps completely around pipe to capture and contain sprays, drips and accumulated condensation.
  • Non-permeable tarp
  • Resists punctures, UV and mildew for long life
  • Quick and easy installation
Non-permeable tarp is vinyl-coated for strength and durability; resists punctures, UV and mildew for long life. Tapered design helps funnel liquid toward center for better drainage. Hook-and-loop fasteners make installation quick and easy and secures diverter in place.