Restek offers a complete line of TO-Can® canisters (SUMMA can equivalent), electropolished using a proprietary process and extensively cleaned using an ultrasonic method.
- Proprietary electropolished surface that maintains compound stability
- High quality, Bellows-design valve
- Now also available with new RAVE™ valve
U.S. EPA Methods TO-14A and TO-15 regulate the collection, storage, and analysis of volatile organic compounds (VOCs) using treated air sampling canisters. This ensures a high-quality, passivated surface that maintains the stability of TO-14A/TO-15 compounds during storage. The frame surrounds the electropolished canister, eliminating the need for weld marks on the sphere, thereby preventing active sites on the canister. The unique holder attaches the handle and base to the canister without welds and protects the canister, tube stub, and valve. Each canister is slightly pressurized with contaminant-free nitrogen prior to shipment.