Dual Canister Sampling Manifold (Stainless Steel & Siltek®-Treated), Restek
Fournisseur: Restek
Certificats
À propos de cet article
Field duplicates of canister samples frequently result in analyte concentrations with high relative standard deviations.
- Duplicate sampling with all canister sizes using one or two flow controllers
- Precise dimensions (9.5" wide x 3.5" high)
- Provides accurate splitting of sample between two canisters
- One-piece design means fewer leaks
- Thick-walled stainless steel tubing is rugged enough for field use
In addition, field duplicates do not differentiate laboratory performance from sampling variability. Restek’s dual canister sampling manifold (DCSM) minimizes sampling variability through a single sample inlet and flow controller by which the sample is evenly collected between two canisters. Use of a single flow controller eliminates flow rate variability, as well as environmental variables common with collocated samples. The DCSM may also be used with two flow controllers to monitor individual canister vacuum.
Ordering information: Do not use the DCSM as a handle to pick up 2 canisters.
Frequently Bought Together