Canister Air Sampling Timer, Restek
Canister Air Sampling Timer, Restek
Canister air sampling timer
  10862-858
 :   Restek
Supplier #:  24267
Canister Air Sampling Timer, Restek
  10862-858
 :  Restek
Canister Air Sampling Timer, Restek
Canister Air Sampling Timer, Restek
Canister air sampling timer
$5,286.77
Each
These timers are designed to simplify both automated and manual air sampling.
  • Program up to 12 timed events
  • Capable of both manual and automated operation
  • Perfect for either grab or time-integrated sampling
  • Long battery life
  • Recharges conveniently using the USB port on any PC
The easy-to-use keypad and graphic display facilitate the programming of up to 12 timed events.
Features include solenoid valve for sampling control, ¹/₄" inlet and outlet fittings, highly inert stainless steel flow path, and waterproof exterior for outdoor use.
They offer the convenience of remote start/stop sampling and permit intermittent sampling throughout a test period. The LCD remains in sleep mode when not in use, greatly extending battery life. Timers are compatible with any canister and flow controller.
  • Désignation:
    Canister air sampling timer