ContactUsLinkComponent
High Visibility Absorbent Mat Pad, PIG®
Catalogue # 10860-054
Fournisseur:  New Pig
undefined
High Visibility Absorbent Mat Pad, PIG®
Catalogue # 10860-054
Fournisseur:  New Pig
Supplier Number:  MAT603
Spécifications
  • Description:
    PIG® High-visibility absorbent mat pad
  • Dimensions:
    15" Wx20" L
  • Cat. no.:
    10860-054
  • Absorbency:
    22 gal./bag
  • Supplier No.:
    MAT603
Spécifications
À propos de cet article
Hi-viz spill absorbents alert workers to potential hazards or repairs.
  • High-visibility yellow color
  • Won't rip, tear or fray even when saturated
  • Absorbs most common industrial liquids
Hi-viz spill absorbents alert workers to potential hazards or repairs. The high-visibility yellow color is easier to see in poorly lit areas to minimize tripping hazards; alerts workers to potential dangers and work in progress. Eight layers of 100% polypropylene are thermally bonded to make PIG Mat the strongest mat on the market; won't rip, tear or fray even when saturated. Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup. Highly absorbent, fine-fiber construction won't leave liquids or fiber residue behind. Absorbs most common industrial liquids - oils, water, solvents, coolants and more.