Kimtech™ A5 Sterile Hood Over Respirator, Kimberly-Clark Professional
Supplier: Ansell Healthcare
Certificates
About this item
Kimtech™ A5 sterile elasticized hoods provide an uninterrupted seal and customized fit for those who wear a full-face respirator.
- Triple bagged, vacuum packed, suitable for ISO Class 5 or higher cleanrooms
- Individually bagged hoods have a sterility indicator and are folded inside out for aseptic donning
- Stretch-fit hood and opening provides an uninterrupted seal around the respirator; great for small faces
- Clean-Don™ technology ties for easy donning and a more secure fit
- Manufactured in an ISO-certified facility, clean-processed and individually packaged under cleanroom conditions
Kimtech™ A5 sterile elasticized hoods feature a stretch fit on the hood and opening, with tunneled overseams to prevent particle shedding. These hoods have an average bacterial filtration efficiency (BFE) of 96% and are made of 100% polypropylene breathable SMS fabric, which gives the wearer cloth-like comfort. This personal protective apparel is triple-bagged and vacuum packaged, suitable for ISO Class 5 or higher cleanrooms. They are made without latex and are designed for single-use (disposable PPE).
Certifications: CE certified.
Specifications
- Catalog No:
- CA10847-814
- 10847-814
- CA75879-282
- Environmentally Preferable:
- Waste Reducing
- Garment application:
- Laboratory
- Laboratory
- Laboratory