To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Size:1 gal., 4/case
- Cat. no.:10017-666
- Packaging:Plastic bottle
- Sterility:Non sterile
- Supplier No.:633508
STERIS's Vesta-Syde® SQ quaternary ammonium disinfectant is a liquid concentrate specifically formulated for disinfection of hard surfaces. The use-dilution can be applied manually or sprayed.
- Flexibility - Excellent fungicidal efficacy can support reduced sporicide use to control mold contamination
- Productivity - Formulated with biodegradable surfactants that provide excellent cleaning ability
- Efficacy - Microbial control against bacteria, fungi and viruses proven using AOAC and EN protocols
- Availability - Active ingredients notified according to the Biocide Product Directive and registered with the US EPA
- Assurance - Manufactured in accordance with the cGMP with complete lot traceability
STERIS' Vesta-Syde® SQ quaternary ammonium disinfectant is the non-sterile version of the Vesta-Syde® SQ family of disinfectants made specifically for use on hard non-porous surfaces.
The Vesta-Syde® SQ products are all made with the same quaternary ammonium formulation, but are processed and packed differently for specific applications. At a use-dilution rate of 0.8% v/v (1:128), the Vesta-Syde® SQ products kill a broad spectrum of common bacteria and fungi without harming the surface.