ContactUsLinkComponent
Micro-organism Lysing VKMix, Precellys® Lysing Kit, Bertin
Catalog # 10144-530
Supplier:  BERTIN CORP
undefined
Micro-organism Lysing VKMix, Precellys® Lysing Kit, Bertin
Catalog # 10144-530
Supplier:  BERTIN CORP
Supplier Number:  P000937-LYSK0-A.0
Specifications
  • Ø:
    0.1 mm and 0.5 mm
  • Description:
    VKMix Micro-organism lysing kit, 7 mL tubes with screw cap and skirt
  • Cat. no.:
    10144-530
  • Supplier No.:
    KT039611304.7
Specifications
About this item
VKMix is made with inert 0.1 mm and 0.5 mm glass beads. It is designed for micro-organism lysing like yeast fungi, gram+, gram- bacteria, spores, micro-algae and more.
  • Ideal for yeast, bacteria and fungi lysis
  • Dnase/Rnase free
  • Autoclavable
Use with the Precellys® Evolution, or Minilys homogenizers.
50 tubes (prefilled)
Certifications: Dnase/Rnase free certified.
Ordering information: Only compatible with Precellys® Evolution and Minilys® homogenizers.
Packaging: Packaged 1 kit contain 50 prefilled and ready-to-use tubes.