Micro-organism Lysing VK01, Precellys® Lysing Kit, Bertin
Supplier: BERTIN CORP
Certificates
About this item
VK01 is made with 0.1 mm inert glass beads and designed for micro-organism lysing like gram-, gram+ bacteria, spores and more.
- Ideal for yeast, bacteria and fungi lysis
- Dnase/Rnase free
- Autoclavable
Use with the Precellys®24, Precellys® Evolution, or Minilys homogenizers.
50 tubes (prefilled)
Certifications: Dnase/Rnase free certified.
Delivery: Packaged 1 kit contain 50 prefilled and ready-to-use tubes.
Frequently Bought Together