ContactUsLinkComponent
Outlet Cap and Gold Seal Assembly Tool for Agilent 1050, 1100 HPLC Systems, Restek
Catalogue # 10853-792
Fournisseur:  Restek
undefined
Outlet Cap and Gold Seal Assembly Tool for Agilent 1050, 1100 HPLC Systems, Restek
Catalogue # 10853-792
Fournisseur:  Restek
Supplier Number:  24989
Spécifications
  • Description:
    Outlet Cap and Gold Seal Assembly Tool
  • Cat. no.:
    10853-792
  • Supplier No.:
    24989
Spécifications
À propos de cet article
Easily install the gold seal into the outlet cap.
Ordering information: For use with Agilent 5001-3701 gold disk seal only. Not compatible with Restek cat.# 25889 and 25890 (Agilent 5067-4728).