ContactUsLinkComponent
Ternary Tee Mixer, Restek
  10853-382
 :  Restek
undefined
Ternary Tee Mixer, Restek
  10853-382
 :  Restek
 :  25138
  • Volume:
    25 µl
  • Description:
    HyperShear Static Mixer, Ternary Tee Mixer 25uL - Stainless Steel
  • Cat. no.:
    10853-382
  • Supplier No.:
    25138
An efficient cross-flow shearing mechanism and interchangeable cartridges produce vortex shear mixing over a wide range of flows.
  • Reduced baseline noise
  • Increased sensitivity
  • Improved gradient accuracy for more reproducible results
  • Increased reaction efficiency in post-column derivatization
Stainless steel.