À propos de cet article
This unique design completely encloses any LC analytical column up to 25 cm in length.
- Easy to set up
- Operation range: 5 °C above ambient to 85 °C, ±1 °C
- Lightweight, compact design fits in small spaces
- Column holder can be placed in any orientation
Two lengths of heater jackets are available: the short column holder accommodates columns up to 10 cm in length, while the long column holder holds columns up to 25 cm in length. The control module provides optimum heating performance, accuracy to within 1 °C, and stability to within 0.1 °C. The new Sidewinder controller has fast 10 Hz sampling for improved responsiveness. The RS232 control allows external programming.
Sidewinder heater units utilize a 24 VDC input to energize the system. This voltage is generated from a universal power supply included with each Sidewinder unit. The power cable supplies current from a wall or bench receptacle to the provided universal 24 VDC universal power supply.