HyperShear Static In-Line Mixers, Restek
Fournisseur: Restek
Certificats
À propos de cet article
An efficient cross-flow shearing mechanism and interchangeable cartridges produce vortex shear mixing over a wide range of flows.
- Reduced baseline noise
- Increased sensitivity
- Improved gradient accuracy for more reproducible results
- Increased reaction efficiency in post-column derivatization
Stainless steel or PEEK.
Frequently Bought Together