LC Piston Seal Insertion Tool, Restek
Catalogue # 10853-846
Fournisseur: Restek
Spécifications
- Description:LC Piston Seal Insertion Tool
- Cat. no.:10853-846
- Supplier No.:21356
Spécifications
À propos de cet article
Simplify pump maintenance.
Use one end to remove your old seal, then simply slip your new seal on the other end and push it flush into position. The tool cannot mar the surrounding metal surface of the pump housing.