Solvent Debubbler, Restek | Avantor
Solvent Debubbler, Restek
Solvent Debubbler With Bracket
Catalog # RK25014
Supplier:   Restek
Fournisseur #:  25014
Solvent Debubbler, Restek
Catalog # RK25014
Supplier:  Restek
Solvent Debubbler, Restek
Solvent Debubbler, Restek
Solvent Debubbler With Bracket
Price
$249.48
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
Bubbles in an LC system can cause check valve malfunctions and pump cavitation, seriously affecting pump performance.
The debubbler removes bubbles from the fluid stream before it enters the pump. Special geometry at the base of the housing allows bubbles entrained in the inlet fluid stream to rise and be trapped in the reservoir. The gas/liquid interface is easily visible through the translucent wall of the device. Loosening the airtight cap releases the trapped gas. The debubbler features a polypropylene body and bracket; universal inlet and outlet connecting tips are made of 316 stainless steel and accept 0.063", 0.085", and 0.125" ID tubing.
Spécifications
  • Désignation:
    Solvent Debubbler With Bracket