Chromium Standards for ICP, Certipur®, MilliporeSigma
- Cr(NO₃)₃ in HNO₃
- Traceable to SRM from NIST
: The quality control of ICP single-element standards is carried out by the accredited calibration laboratory according to ISO 17025.
: Includes a Certificate of Analysis, with exact data on content including uncertainty budget, trace element impurities, composition, traceability, date of release and minimum shelf life.
- Hazard class description:Corrosive Material
Frequently Bought Together