About this item
The new "Advanced Technology" foam is manufactured specifically for the controlled environment industry. This foam allows 25% more liquid absorption and two times more liquid release than competing foams. Each refill is completely autoclavable and resistant to gamma and ETO sterilization. Refills are individually packaged and lot number controlled. All Vileda® Professional brand refills are ideal for use with the "Pull and Lift" technique most commonly used in controlled environments. Manufactured in the USA.
- Original refills have been the standard in the professional and controlled environment industry for well over 30 years
- Universal refills are perfect for general use within all controlled environments
- Designed for use in all Roll-O-Matic® applications
- Fully autoclavable
- Manufactured and sterilized in the USA
- Validated to 10¯⁶ SAL per ISO 11137
- Individually double bagged with Lot # and indicator Dot on each package
- Certificate of Sterility available upon request with each shipment
The Roll-O-Matic® "Disinfect" Refill features polyester lamination that allows for a tear-free application of disinfectants, cleaning, and spill absorption
The Roll-O-Matic® "Clean" Refill features microfiber lamination that allows for a tear-free deep cleaning of contamination and residues, while also applying disinfectants to the surfaces. It also evenly distributes disinfectants to optimize manufacturer recommended contact times. Unlike competitive products, this microfiber is laminated to the foam without adhesives, which could contaminate surfaces. The Stainless Steel channel allows for multiple uses with multiple autoclave cycles.
All refill styles are ideal for use in ISO 5, LAR, production, and laboratory environments.
It evenly distributes disinfectants to optimize manufacturer recommended contact times. Unlike competitive products, this polyester is laminated to the foam without adhesives, which could contaminate surfaces. The Stainless Steel channel allows for multiple uses with multiple autoclave cycles.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.