WaterPro® RO Systems Accessories, Labconco
: Labconco
Prefilter/Carbon Filter Kit required for operation of the WaterPro® RO System.
: WaterPro® RO System sold separately.
: Kit includes Prefilters (3) and Carbon Filters (3) for initial start-up, and two filter changes. The Reverse Osmosis Membrane is required for operation of WaterPro® RO System.
Frequently Bought Together