Rhodium Standards for ICP, Certipur®, MilliporeSigma
Catalog # CA1.70345.0100
Supplier: MilliporeSigma
CAS Number:
Specifications
- Concentration:1000 µg/ml
- Matrix:2-3 % HNO₃
- Pack type:Plastic Bottle
- Size:100 ml
- Cat. no.:CA1.70345.0100
- Element:Rhodium
- Supplier No.:1.70345.0100
Specifications
About this item
Rh(NO₃)₃ in HNO₃
Traceable to SRM from NIST.
Certifications: The quality control of ICP single-element standards is carried out by the accredited calibration laboratory according to ISO 17025.
Ordering information: Includes a Certificate of Analysis, with exact data on content including uncertainty budget, trace element impurities, composition, traceability, date of release and minimum shelf life.