Antimony Standard for ICP, Certipur®, MilliporeSigma
Catalogue # CA1.70302.0100
Fournisseur: MilliporeSigma
Numéro CAS:
Spécifications
- Concentration:1000 µg/ml
- Matrix:7% HCl
- Pack type:Plastic Bottle
- Size:100 ml
- Cat. no.:CA1.70302.0100
- Element:Antimony
- Supplier No.:1.70302.0100
Spécifications
À propos de cet article
Traceable to SRM from NIST.
- Sb₂O₃ in HCl
Certifications: The quality control of ICP single-element standards is carried out by the accredited calibration laboratory according to ISO 17025.
Ordering information: Includes a Certificate of Analysis, with exact data on content including uncertainty budget, trace element impurities, composition, traceability, date of release and minimum shelf life.