TSK-GEL® Cation Exchange Chromatography Columns, Tosoh Bioscience
New Product
Supplier:   Tosoh Bioscience
TSK-GEL® Cation Exchange Chromatography Columns, Tosoh Bioscience
Supplier:  Tosoh Bioscience
New Product
Certificates
TSK-GEL® Cation Exchange Chromatography Columns, Tosoh Bioscience
TSK-GEL® Cation Exchange Chromatography Columns, Tosoh Bioscience
24 Options Available Below
Product Details & Documents
Efficient chromatography can be achieved with the nonporous TSKgel STAT columns due to the novel bonding chemistry and the absence of micro-pores. Applications for the TSKgel CM-STAT and TSKgel SP-STAT columns include the separation of proteins, protein aggregates,charge variants of monoclonal antibodies, PEGylated proteins, and peptide digests.
The pore structure and bonding chemistry of TSKgel BioAssist S columns provides high capacity for medium to large molar mass proteins.
TSKgel CM-5PW and TSKgel SP-5PW columns are an excellent choice for analyzing biologically active molecules.
TSKgel SP-NPR nonporous columns are used for the separation and analysis of proteins and peptides. They are particularly useful for adeno-associated viruses and other large macromolecules.
TSKgel CM-2SW and TSKgel SP-2SW (both 5 µm, 12.5 nm) and TSKgel CM-3SW (10 µm, 25 nm) are available for analyzing smaller molar mass samples such as nucleotides, drug candidates, catecholamines, and small peptides or proteins
TSKgel SCX columns are used for the separation and analysis of organic acids, saccharides and alcohols.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 78048-718
  • 78048-730
  • 78048-768
  • 78049-182
  • 78048-524
  • 78048-528
  • 78048-530
  • 78048-604
  • 78048-736
  • 78048-744
  • 78049-072
  • 78049-414
  • 78049-408
  • 78049-410
  • 78049-412
  • 78048-538
  • 78048-542
  • 78048-532
  • 78048-550
  • 78048-616
  • 78048-726
  • 78048-738
  • 78048-832
  • 78048-610
  • Max. pressure:
  • 10 bar
  • 15 bar
  • 15 bar
  • 50 bar
  • 50 bar
  • 15 bar
  • 25 bar
  • 15 bar
  • 200 bar
  • 10 bar
  • 100 bar
  • 100 bar
  • 100 bar
  • 100 bar
  • 150 bar
  • 150 bar
  • 25 bar
  • pH range:
  • 2.0 - 12.0
  • 2.0 - 12.0
  • 2.0 - 12.0
  • 2.0 - 12.0
  • 2.0 - 12.0
  • 2.0 - 12.0
  • 2.0 - 12.0
  • 2.0 - 12.0
  • 2.0 - 12.0
  • 2.0 - 12.0
  • 2.0 - 12.0
  • 3.0 - 10.0
  • 3.0 - 10.0
  • 3.0 - 10.0
  • 3.0 - 10.0
  • 2.0 - 7.5
  • 2.0 - 7.5
  • 2.0 - 7.5
  • 2.0 - 12.0
  • 2.0 - 7.5
  • 2.0 - 12.0
  • 2.0 - 12.0
  • 2.0 - 12.0
  • 2.0 - 7.5
  • Temperature limit:
  • 45 °C
  • 45 °C
  • 45 °C
  • 60 °C
  • 45 °C
  • 45 °C
  • 45 °C
  • 45 °C
  • 60 °C
  • 45 °C
  • 60 °C
  • 45 °C
  • 60 °C
  • 30 °C
  • 45 °C
  • 45 °C
  • 60 °C
  • 45 °C
  • 45 °C
  • 45 °C
  • 45 °C
  • 45 °C
  • 45 °C
  • USP code:
  • L58
  • L22
  • L9
Spécifications
Product Family Options
Polymer based glass columns
Informations sur le produit
  • Désignation
    Type de colonne
    Phase
    Taille de pore
    Column Hardware
    Granulométrie
    Ø int.
    Longueur
    Availability
    Price
  • TSK-GEL® SP-5PW
    Analytique
    Sulfopropyl
    1000 Å
    Verre
    10 µm
    8.00 mm
    75 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,788.57
    $3,788.57
    Each
       $3,788.57
    Supplier Number: 08803
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78048-718
    Max. pressure
    10 bar
    pH range
    2.0 - 12.0
    Temperature limit
    45 °C
    USP code
    New Product
  • TSK-GEL® SP-5PW
    Analytique
    Sulfopropyl
    1000 Å
    Verre
    10 µm
    5.00 mm
    50 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,906.74
    $2,906.74
    Each
       $2,906.74
    Supplier Number: 13062
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78048-730
    Max. pressure
    15 bar
    pH range
    2.0 - 12.0
    Temperature limit
    45 °C
    USP code
    New Product
  • TSK-GEL® SP-5PW
    Semi-préparative
    Sulfopropyl
    1000 Å
    Verre
    13 µm
    20.00 mm
    150 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $16,264.70
    $16,264.70
    Each
       $16,264.70
    Supplier Number: 14017
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78048-768
    Max. pressure
    15 bar
    pH range
    2.0 - 12.0
    Temperature limit
    45 °C
    USP code
    New Product
Polymer based PEEK columns
Informations sur le produit
  • Désignation
    Type de colonne
    Phase
    Taille de pore
    Column Hardware
    Granulométrie
    Ø int.
    Longueur
    Availability
    Price
  • TSK-GEL® BioAssist S
    Analytique
    Sulfopropyl
    1300 Å
    PEEK
    7 µm
    4.60 mm
    50 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,347.64
    $3,347.64
    Each
       $3,347.64
    Supplier Number: 19686
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78049-182
    Max. pressure
    pH range
    2.0 - 12.0
    Temperature limit
    USP code
    New Product
Polymer based stainless steel columns
Informations sur le produit
  • Désignation
    Type de colonne
    Phase
    Taille de pore
    Column Hardware
    Granulométrie
    Ø int.
    Longueur
    Availability
    Price
  • TSK-GEL® SCX Na+
    Analytique
    Sulfonic Acid
    60 Å
    Acier inoxydable
    5 µm
    6.00 mm
    150 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,993.85
    $2,993.85
    Each
       $2,993.85
    Supplier Number: 07156
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78048-524
    Max. pressure
    50 bar
    pH range
    2.0 - 12.0
    Temperature limit
    60 °C
    USP code
    L58
    New Product
  • TSK-GEL® SCX H+
    Analytique
    Sulfonic Acid
    60 Å
    Acier inoxydable
    5 µm
    7.80 mm
    300 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,720.52
    $3,720.52
    Each
       $3,720.52
    Supplier Number: 07158
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78048-528
    Max. pressure
    50 bar
    pH range
    2.0 - 12.0
    Temperature limit
    45 °C
    USP code
    L22
    New Product
  • TSK-GEL® SP-5PW
    Analytique
    Sulfopropyl
    1000 Å
    Acier inoxydable
    10 µm
    7.50 mm
    75 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,704.18
    $3,704.18
    Each
       $3,704.18
    Supplier Number: 07161
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78048-530
    Max. pressure
    15 bar
    pH range
    2.0 - 12.0
    Temperature limit
    45 °C
    USP code
    New Product
  • TSK-GEL® SP-5PW
    Semi-préparative
    Sulfopropyl
    1000 Å
    Acier inoxydable
    13 µm
    21.50 mm
    150 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $12,862.63
    $12,862.63
    Each
       $12,862.63
    Supplier Number: 07575
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78048-604
    Max. pressure
    25 bar
    pH range
    2.0 - 12.0
    Temperature limit
    45 °C
    USP code
    New Product
  • TSK-GEL® CM-5PW
    Analytique
    Carboxymethyl
    1000 Å
    Acier inoxydable
    10 µm
    7.50 mm
    75 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,704.18
    $3,704.18
    Each
       $3,704.18
    Supplier Number: 13068
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78048-736
    Max. pressure
    15 bar
    pH range
    2.0 - 12.0
    Temperature limit
    45 °C
    USP code
    New Product
  • TSK-GEL® SP-NPR, non porous
    Analytique
    Sulfopropyl
    1000 Å
    Acier inoxydable
    2.5 µm
    4.60 mm
    35 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,874.07
    $2,874.07
    Each
       $2,874.07
    Supplier Number: 13076
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78048-744
    Max. pressure
    200 bar
    pH range
    2.0 - 12.0
    Temperature limit
    60 °C
    USP code
    New Product
  • TSK-GEL® SP-5PW
    Analytique
    Sulfopropyl
    1000 Å
    Acier inoxydable
    10 µm
    2.00 mm
    75 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,789.71
    $2,789.71
    Each
       $2,789.71
    Supplier Number: 18758
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78049-072
    Max. pressure
    10 bar
    pH range
    2.0 - 12.0
    Temperature limit
    45 °C
    USP code
    New Product
  • CM-STAT, non porous
    Analytique
    Carboxymethyl
    Acier inoxydable
    7 µm
    4.60 mm
    100 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,994.41
    $2,994.41
    Each
       $2,994.41
    Supplier Number: 21966
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78049-414
    Max. pressure
    100 bar
    pH range
    3.0 - 10.0
    Temperature limit
    60 °C
    USP code
    New Product
  • SP-STAT, non porous
    Analytique
    Sulfopropyl
    Acier inoxydable
    10 µm
    3.00 mm
    35 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,830.92
    $2,830.92
    Each
       $2,830.92
    Supplier Number: 21963
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78049-408
    Max. pressure
    100 bar
    pH range
    3.0 - 10.0
    Temperature limit
    45 °C
    USP code
    New Product
  • SP-STAT, non porous
    Analytique
    Sulfopropyl
    Acier inoxydable
    7 µm
    4.60 mm
    100 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,994.41
    $2,994.41
    Each
       $2,994.41
    Supplier Number: 21964
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78049-410
    Max. pressure
    100 bar
    pH range
    3.0 - 10.0
    Temperature limit
    60 °C
    USP code
    New Product
  • CM-STAT, non porous
    Analytique
    Carboxymethyl
    Acier inoxydable
    10 µm
    3.00 mm
    35 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,830.92
    $2,830.92
    Each
       $2,830.92
    Supplier Number: 21965
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78049-412
    Max. pressure
    100 bar
    pH range
    3.0 - 10.0
    Temperature limit
    30 °C
    USP code
    New Product
Silica based stainless steel columns
Informations sur le produit
  • Désignation
    Type de colonne
    Phase
    Taille de pore
    Column Hardware
    Granulométrie
    Ø int.
    Longueur
    Availability
    Price
  • TSK-GEL® SP-2SW
    Analytique
    Sulfopropyl
    125 Å
    Acier inoxydable
    5 µm
    4.60 mm
    250 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,908.31
    $3,908.31
    Each
       $3,908.31
    Supplier Number: 07165
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78048-538
    Max. pressure
    150 bar
    pH range
    2.0 - 7.5
    Temperature limit
    45 °C
    USP code
    L9
    New Product
  • TSK-GEL® CM-2SW
    Analytique
    Carboxymethyl
    125 Å
    Acier inoxydable
    5 µm
    4.60 mm
    250 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,347.64
    $3,347.64
    Each
       $3,347.64
    Supplier Number: 07167
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78048-542
    Max. pressure
    150 bar
    pH range
    2.0 - 7.5
    Temperature limit
    45 °C
    USP code
    New Product
  • TSK-GEL® CM-3SW
    Analytique
    Carboxymethyl
    250 Å
    Acier inoxydable
    10 µm
    7.50 mm
    75 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,347.64
    $3,347.64
    Each
       $3,347.64
    Supplier Number: 07162
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78048-532
    Max. pressure
    25 bar
    pH range
    2.0 - 7.5
    Temperature limit
    60 °C
    USP code
    New Product
Guard column
Informations sur le produit
  • Désignation
    Type de colonne
    Phase
    Taille de pore
    Column Hardware
    Granulométrie
    Ø int.
    Longueur
    Availability
    Price
  • TSK-GEL® SP-5PW Guardgel Kit
    Colonne de garde
    Sulfopropyl
    1000 Å
    Acier inoxydable
    20 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,725.53
    $1,725.53
    Each
       $1,725.53
    Supplier Number: 07211
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78048-550
    Max. pressure
    pH range
    2.0 - 12.0
    Temperature limit
    45 °C
    USP code
    New Product
  • TSK-GEL® CM-SW Guardgel Kit
    Colonne de garde
    Carboxymethyl
    125 Å
    Acier inoxydable
    10 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,793.58
    $1,793.58
    Each
       $1,793.58
    Supplier Number: 07650
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78048-616
    Max. pressure
    pH range
    2.0 - 7.5
    Temperature limit
    45 °C
    USP code
    New Product
  • TSK-GEL® SP-5PW Guardgel Kit, Glass
    Colonne de garde
    Sulfopropyl
    1000 Å
    Verre
    20 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,839.85
    $1,839.85
    Each
       $1,839.85
    Supplier Number: 08807
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78048-726
    Max. pressure
    pH range
    2.0 - 12.0
    Temperature limit
    45 °C
    USP code
    New Product
  • TSK-GEL® CM-5PW Guardgel Kit
    Colonne de garde
    Carboxymethyl
    1000 Å
    Acier inoxydable
    10 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,725.53
    $1,725.53
    Each
       $1,725.53
    Supplier Number: 13069
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78048-738
    Max. pressure
    pH range
    2.0 - 12.0
    Temperature limit
    45 °C
    USP code
    New Product
  • TSK-GEL® SP-5PW, Prep Guardgel Kit
    Colonne de garde
    Sulfopropyl
    1000 Å
    Acier inoxydable
    20 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,653.61
    $2,653.61
    Each
       $2,653.61
    Supplier Number: 16093
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78048-832
    Max. pressure
    pH range
    2.0 - 12.0
    Temperature limit
    45 °C
    USP code
    New Product
Informations sur le produit
  • Désignation
    Type de colonne
    Phase
    Taille de pore
    Column Hardware
    Granulométrie
    Ø int.
    Longueur
    Availability
    Price
  • TSKgel SP-2SW Guardgel kit
    Colonne de garde
    Sulfopropyl
    125 Å
    Acier inoxydable
    5 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,575.83
    $1,575.83
    Each
       $1,575.83
    Supplier Number: 07644
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78048-610
    Max. pressure
    pH range
    2.0 - 7.5
    Temperature limit
    45 °C
    USP code
    New Product