TSK-Gel® Reversed Phase Columns, Tosoh Bioscience
TSK-Gel® Reversed Phase Columns, Tosoh Bioscience
New Product
Supplier:   Tosoh Bioscience
TSK-Gel® Reversed Phase Columns, Tosoh Bioscience
Supplier:  Tosoh Bioscience
New Product
Certificates
TSK-Gel® Reversed Phase Columns, Tosoh Bioscience
TSK-Gel® Reversed Phase Columns, Tosoh Bioscience
10 Options Available Below
Product Details & Documents
Recommended for small molecular weight compounds (<10000 Da) such as peptides, amino acids, tryptic digests, nucleotides, pharmaceutical molecules, and food and beverage samples.
  • Provide superior resolution and speed
  • Improved sensitivity
  • Pressure drop is not excessive due to the monodisperse particle size distribution
TSKgel® Super-ODS, Super-Octyl and Super-Phenyl packings are bonded with, respectively, C18, C8 and phenyl functional groups. The bonded phases have a polymeric structure. An exhaustive endcapping reaction minimizes the presence of residual silanol groups.
Product Family Options
Informations sur le produit
  • Désignation
    Phase
    Taille de pore
    Column Hardware
    Granulométrie
    Ø int.
    Longueur
    Débit
    Packing Material
    Availability
    Price
  • TSK-Gel® Super-ODS, 11 nm
    C18 (ODS or Octadecyl)
    110 Å
    Acier inoxydable
    2 µm
    4.60 mm
    50 mm
    1 - 2.5 ml/min
    X, e
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 627,55 $
    1 627,55 $
    Each
       1 627,55 $
    Supplier Number: 18154
    Spécifications:
    New Product
    New Product
  • TSK-Gel® Super-ODS, 11 nm
    C18 (ODS or Octadecyl)
    110 Å
    Acier inoxydable
    2 µm
    4.60 mm
    100 mm
    1 - 2.5 ml/min
    Mmid, e
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 692,88 $
    1 692,88 $
    Each
       1 692,88 $
    Supplier Number: 18197
    Spécifications:
    New Product
    New Product
  • TSK-Gel® Super-Octyl, 11 nm
    C8 (Octyl)
    110 Å
    Acier inoxydable
    2 µm
    4.60 mm
    50 mm
    1 - 2.5 ml/min
    X, e
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 627,55 $
    1 627,55 $
    Each
       1 627,55 $
    Supplier Number: 18275
    Spécifications:
    New Product
    New Product
  • TSK-Gel® Super-Octyl, 11 nm
    C8 (Octyl)
    110 Å
    Acier inoxydable
    2 µm
    4.60 mm
    100 mm
    1 - 2.5 ml/min
    Mm, e
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 692,88 $
    1 692,88 $
    Each
       1 692,88 $
    Supplier Number: 18276
    Spécifications:
    New Product
    New Product
  • TSK-Gel® Super-Phenyl, 11 nm
    Phenyl
    110 Å
    Acier inoxydable
    2 µm
    4.60 mm
    50 mm
    1 - 2.5 ml/min
    Mm, e
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 627,55 $
    1 627,55 $
    Each
       1 627,55 $
    Supplier Number: 18277
    Spécifications:
    New Product
    New Product
  • TSK-Gel® Super-Phenyl, 11 nm
    Phenyl
    110 Å
    Acier inoxydable
    2 µm
    4.60 mm
    100 mm
    1 - 2.5 ml/min
    Mm, e
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 692,88 $
    1 692,88 $
    Each
       1 692,88 $
    Supplier Number: 18278
    Spécifications:
    New Product
    New Product
  • TSK-Gel® Super-ODS, 11 nm
    C18 (ODS or Octadecyl)
    110 Å
    Acier inoxydable
    2 µm
    2.00 mm
    50 mm
    0.15 - 0.2 ml/min
    Slumns, Tosohoel® ReverroKyl, -
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 529,58 $
    1 529,58 $
    Each
       1 529,58 $
    Supplier Number: 19541
    Spécifications:
    New Product
    New Product
  • TSK-Gel® Super-ODS, 11 nm
    C18 (ODS or Octadecyl)
    110 Å
    Acier inoxydable
    2 µm
    2.00 mm
    100 mm
    0.15 - 0.2 ml/min
    Slumns, Tosohoel® ReverroKyl, -
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 573,12 $
    1 573,12 $
    Each
       1 573,12 $
    Supplier Number: 19542
    Spécifications:
    New Product
    New Product
  • TSK-Gel® Super-Octyl, 11 nm
    C8 (Octyl)
    110 Å
    Acier inoxydable
    2 µm
    2.00 mm
    50 mm
    0.15 - 0.20 ml/min
    Slumns, Tosohoel® ReverroKyl, -
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 529,58 $
    1 529,58 $
    Each
       1 529,58 $
    Supplier Number: 20013
    Spécifications:
    New Product
    New Product
  • TSK-Gel® Super-Octyl, 11 nm
    C8 (Octyl)
    110 Å
    Acier inoxydable
    2 µm
    2.00 mm
    100 mm
    0.15 - 0.20 ml/min
    Slumns, Tosohoel® ReverroKyl, -
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 657,84 $
    1 657,84 $
    Each
       1 657,84 $
    Supplier Number: 20014
    Spécifications:
    New Product