Lab Guard® UV Protection Bags
Fournisseur: Minigrip
Certificats
À propos de cet article
LAB GUARD® UV Protection bag contains UV blocker film to prevent bag contents from deterioration.
- Premium zipper that opens quickly and easily
- Amber color makes identification of light-sensitive contents easy
- Available in convenient dispenser bag
Complies with USP XXII and USP XXIII procedure 66.1 with light transmission not exceeding 10% at any wavelength from 290 to 450 nm.
Frequently Bought Together