CTAB Extraction Solution, Teknova | Avantor
CTAB Extraction Solution, Teknova
Supplier:   Teknova
CTAB Extraction Solution, Teknova
Supplier:  Teknova
Certificates
CTAB Extraction Solution, Teknova
1 Options Available Below
Product Details & Documents
CTAB is a cationic detergent used in the extraction of genomic DNA from bacteria and for the isolation of high molecular weight DNA from plants for plamid minipreps and for PCR.
The Solution contains 1% CTAB, 100 mM Tris-HCl (pH 8,0), 20mM EDTA (pH 8,0) and 1.4M Sodium Chloride.
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Availability
    Price
  • CTAB extraction solution, sterile
    250 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $132.40
    $132.40
    Each
       $132.40
    La quantité minimale de commande est de 4
    Supplier Number: C2190
    Spécifications:
    New Product
    New Product
  • CTAB extraction solution, sterile
    250 ml
    Discontinued
    Supplier Number: -