PIE-Blocks, Reaction Blocks, Chemglass
Catalog # 17006-592
Supplier: Chemglass
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Description:PIE-BLOCK System
- Cat. no.:17006-592
- Supplier No.:CG-1991-P-01
Specifications
About this item
Reaction PIE-BLOCK is designed to fit directly on 135 mm diameter circular top hot plate stirrers
- Holes are spaced for optimum stirring of all vials
- Inserts are color coded for quick and easy identification
- Interchangeable pie wedges permit various sized vials on one hot plate
- Aluminum construction provides excellent heat transfer
- Safety pie plate holder keeps up to four pie wedges securely locked together on hotplate
A drilled thermowell is provided for insertion of contact thermometers or temperature sensors.
Ordering information: Supplied with safety Holder and 6 listed Pie Wedges.