TSK-Gel® NPH Columns, Tosoh Bioscience
New Product
Supplier: Tosoh Bioscience
À propos de cet article
TSK-Gel® NPH columns are stainless steel with polymer based.
7 Options Available Below
Product Details & Documents
TSK-Gel® NPH columns are stainless steel with polymer based.
Specifications for Product Family
Spécifications
- Catalog No:
- 78048-878
- 78048-994
- 78048-526
- 78048-742
- 78048-822
- 78048-992
- 78049-692
- Max. pressure:
- 200 bar
- pH range:
- 1.0 - 14.0
- 2.0 - 12.0
- 2.0 - 12.0
- 2.0 - 12.0
- 2.0 - 12.0
- Temperature limit:
- 60 °C
Spécifications
Product Family Options
Guard columns
Informations sur le produit
- DésignationType de colonnePhaseColumn HardwareGranulométrieØ int.LongueurPacking MaterialAvailabilityPrice
- TSK-Gel® DEAE-NPR Guard columnColonne de gardeDEAEAcier inoxydable5 µm4.60 mm5 mmSilicaLog in pour la disponibilitéEach$985.25Supplier Number: 17088Spécifications:New Product
More
Specifications
Cat. No.78048-878Max. pressurepH rangeTemperature limitNew Product - TSK-Gel® DNA-NPR Guard columnColonne de gardeDEAEAcier inoxydable5 µm4.60 mm5 mmSilicaLog in pour la disponibilitéEach$985.25Supplier Number: 18253Spécifications:New Product
More
Specifications
Cat. No.78048-994Max. pressurepH rangeTemperature limitNew Product
Informations sur le produit
- DésignationType de colonnePhaseColumn HardwareGranulométrieØ int.LongueurPacking MaterialAvailabilityPrice
- TSK-Gel® SAXAnalytiqueSAX (Strong Anion Exchange)Acier inoxydable5 µm6.00 mm150 mmSilicaLog in pour la disponibilitéEach$2,841.43Supplier Number: 07157Spécifications:New Product
More
Specifications
Cat. No.78048-526Max. pressurepH range1.0 - 14.0Temperature limitNew Product - TSK-Gel® DEAE-NPR, non porousAnalytiqueDEAEAcier inoxydable2.5 µm4.60 mm35 mmSilicaLog in pour la disponibilitéEach$2,874.07Supplier Number: 13075Spécifications:New Product
More
Specifications
Cat. No.78048-742Max. pressurepH range2.0 - 12.0Temperature limitNew Product - TSK-Gel® Butyl-NPR, non porousAnalytiqueButylAcier inoxydable2.5 µm4.60 mm35 mmSilicaLog in pour la disponibilitéEach$3,378.66Supplier Number: 14947Spécifications:New Product
More
Specifications
Cat. No.78048-822Max. pressurepH range2.0 - 12.0Temperature limitNew Product - TSK-Gel® DNA-NPR, non porousAnalytiqueDEAEAcier inoxydable2.5 µm4.60 mm75 mmSilicaLog in pour la disponibilitéEach$3,908.31Supplier Number: 18249Spécifications:New Product
More
Specifications
Cat. No.78048-992Max. pressurepH range2.0 - 12.0Temperature limitNew Product - TSKgel Butyl-NPRAnalytiqueButylAcier inoxydable2.5 µm4.60 mm100 mmSilicaLog in pour la disponibilitéEach$3,771.71Supplier Number: 42168Spécifications:New Product
More
Specifications
Cat. No.78049-692Max. pressure200 barpH range2.0 - 12.0Temperature limit60 °CNew Product