À propos de cet article
The less time spent on cleanroom cleaning, the less interference with keeping the core business running. The unique CE Duo Mop concept is the perfect answer for the high demands of effective cleaning and disinfection of cleanrooms and controlled environments. It is faster, easier, and twice as efficient as traditional methods. The CE Duo Mops allow users to achieve contact time requirements for common disinfectants, and still reach an excellent cleaning performance, with up to 99.99% bacteria removal. The CleanTech™ and SanTech™ Duo Mops form the most efficient and reliable cleaning concept in the market for contamination control.
- Designed for use in cleanrooms up to close ISO 5/GMP A&B/FED209D Class 100
- Adhesive-free lamination techniques are used to prevent additional product contamination
- Suitable for bucket and pre-prepared method
- Sterile items are validated to 10-6 SAL per ISO 11137
- Manufactured and sterilized in the USA
- Certificate of Sterility available upon request with each shipment
CleanTech™ Mops feature microfiber laminated cleanroom foam for cleaning/disinfecting floors, walls, and ceilings
SanTech™ Mops feature polyester laminated cleanroom foam for disinfecting floors, walls, and ceilings. These mops offer excellent liquid absorbency. They are individually double bagged with Lot Number and Indicator Dot on each package.
Excellent microfiber cleaning results have been independently proven to provide bacteria reduction of 99.997%. CleanTech™ Mops are individually double bagged with Lot Number and Indicator Dot on each package.