ContactUsLinkComponent
Puritan® Mini-Tip Foam Tipped Applicator, Wooden Handle, Puritan Medical Products
Puritan® Mini-Tip Foam Tipped Applicator, Wooden Handle, Puritan Medical Products
Catalog # 89133-778
Product available on GSA Advantage®
undefined
Puritan® Mini-Tip Foam Tipped Applicator, Wooden Handle, Puritan Medical Products
Catalog # 89133-778
Supplier Number:  1803-WF
Product available on GSA Advantage®
Specifications
  • Cat. no.:
    89133-778
  • Packaging:
    50 Swabs/Bag, 20 Bags/Case
  • Handle Length:
    74.6 mm (2.937")
  • Head Width:
    4 mm (0.157")
  • Head Length:
    12 mm (0.472")
  • Handle Width:
    2.1 mm (0.827")
  • Supplier No.:
    1803-WF
Specifications
About this item
This specialty foam tipped applicator is produced with 100ppi reticulated medical grade foam
Made in the USA.
It has a soft, non-abrasive tip that is bonded securely to a white birch handle with thermoplastic adhesive.
Certifications: CE marked.