TSK-Gel® Reversed Phase Columns, Tosoh Bioscience
New Product
TSK-Gel® Reversed Phase Columns, Tosoh Bioscience
New Product
TSK-Gel® Reversed Phase Columns, Tosoh Bioscience
TSK-Gel® Reversed Phase Columns, Tosoh Bioscience
These silica- and polymer-based columns are ideal for reversed-phase applications such as pharmaceutical drugs, forensic compounds, derivatised amino acids, carbohydrates, steroids, lipids and fatty acids. Depending on their molecular weight, peptides may be analysed with the 80 to 140 Å silica-based columns, while proteins require a larger pore size column (200 to 300 Å).
  • Silica and polymer-based materials
  • Super series ideal for LCMS
  • High recovery for biomolecules
TSKgel® Protein C4-300:
TSKgel® Protein C4-300 consists of a polymerically bonded butyl (C4) phase on 300 Å ultra-pure silica. The latest surface modification techniques and endcapping of residual silanol groups, reduce undesirable secondary interactions and peak tailing. It has been designed for the optimal recovery and resolution of biomolecules such as recombinant proteins, antibody fragments or PEGylated proteins.
TSKgel® ODS-140HTP:
TSKgel® ODS-140HTP columns were developed for use in high throughput (HTP) applications including drug discovery, pharmacokinetics and peptide digest separations. They are packed with 2.3 µm particles, providing high resolution and short analysis times at moderate pressure. The lower pressure drop enables these columns to be used with either UHPLC (up to 9006.84 psi) or conventional HPLC systems.
TSKgel® Super series:
The silica particles used in the Super series columns are monodisperse spherical 2.3 µm beads which are polymerically bonded and exhaustively end capped, to produce low metal-content, low silanol phases. The 2.3 µm particle provides superior resolution and speed of analysis as well as improved sensitivity. The columns do not suffer from excessive pressure drops.
Polymer-based phases:
The polymer-based reversed phase materials are chemically stable from pH 2 to 12, allowing operation at basic pH where silica-based phases have limited chemical stability. The non-porous Octadecyl-NPR column produces extremely fast kinetics and quantitative recovery of proteins at sub-microgram loads. TSKgel® Octadecyl-4PW is suitable for the analysis of peptides and small proteins, whereas the TSKgel® Octadecyl-2PW is used for small pharmaceutical compounds at basic pH. TSKgel® Phenyl-5PW has a high loading capacity and is ideal for the separation of high molecular weight proteins.
Product Family Options
Silica
  • Désignation
    Phase
    Taille de pore
    Column Hardware
    Granulométrie
    Ø int.
    Longueur
    Packing Material
  • TSKgel ODS-120T
    C18 (ODS or Octadecyl)
    120 Å
    Acier inoxydable
    5 µm
    4.60 mm
    250 mm
    Silica
    Each
    Each - $1,442.49
    $1,442.49
    Each
       $1,442.49
     : 07125
    New Product
    New Product
  • TSKgel TMS-250
    250 Å
    Acier inoxydable
    10 µm
    4.60 mm
    75 mm
    Silica
    Each
    Each - $2,174.62
    $2,174.62
    Each
       $2,174.62
     : 07190
    New Product
    New Product
  • TSKgel ODS-120T
    C18 (ODS or Octadecyl)
    120 Å
    Acier inoxydable
    5 µm
    4.60 mm
    150 mm
    Silica
    Each
    Each - $1,368.99
    $1,368.99
    Each
       $1,368.99
     : 07637
    New Product
    New Product
  • TSKgel ODS-80TS
    C18 (ODS or Octadecyl)
    80 Å
    Acier inoxydable
    5 µm
    2.00 mm
    150 mm
    Silica
    Each
    Each - $1,268.29
    $1,268.29
    Each
       $1,268.29
     : 18150
    New Product
    New Product
  • TSKgel ODS-80TS
    C18 (ODS or Octadecyl)
    80 Å
    Acier inoxydable
    5 µm
    2.00 mm
    250 mm
    Silica
    Each
    Each - $1,368.99
    $1,368.99
    Each
       $1,368.99
     : 18151
    New Product
    New Product
  • TSKgel ODS-80TS
    C18 (ODS or Octadecyl)
    80 Å
    Acier inoxydable
    5 µm
    4.60 mm
    150 mm
    Silica
    Each
    Each - $1,368.99
    $1,368.99
    Each
       $1,368.99
     : 17201
    New Product
    New Product
  • TSKgel ODS-80TS
    C18 (ODS or Octadecyl)
    80 Å
    Acier inoxydable
    5 µm
    4.60 mm
    250 mm
    Silica
    Each
    Each - $1,442.49
    $1,442.49
    Each
       $1,442.49
     : 17202
    New Product
    New Product
Polymer
  • Désignation
    Phase
    Taille de pore
    Column Hardware
    Granulométrie
    Ø int.
    Longueur
    Packing Material
  • TSKgel Octadecyl-NPR
    C18 (ODS or Octadecyl)
    Acier inoxydable
    2.5 µm
    4.60 mm
    35 mm
    Silica
    Each
    Each - $2,640.03
    $2,640.03
    Each
       $2,640.03
     : 14005
    New Product
    New Product
  • TSKgel Phenyl-5PW
    Phenyl
    1000 Å
    Acier inoxydable
    10 µm
    4.60 mm
    75 mm
    Silica
    Each
    Each - $1,788.14
    $1,788.14
    Each
       $1,788.14
     : 08043
    New Product
    New Product
  • TSKgel Octadecyl-4PW
    C18 (ODS or Octadecyl)
    500 Å
    Acier inoxydable
    7 µm
    4.60 mm
    150 mm
    Silica
    Each
    Each - $1,788.14
    $1,788.14
    Each
       $1,788.14
     : 13351
    New Product
    New Product
  • TSKgel Octadecyl-2PW
    C18 (ODS or Octadecyl)
    100 - 200 Å
    Acier inoxydable
    5 µm
    4.60 mm
    150 mm
    Silica
    Each
    Each - $2,003.13
    $2,003.13
    Each
       $2,003.13
     : 17500
    New Product
    New Product
Guard columns
  • Désignation
    Phase
    Taille de pore
    Column Hardware
    Granulométrie
    Ø int.
    Longueur
    Packing Material
  • Guard filter holder
    C18 (ODS or Octadecyl)
    Acier inoxydable
    4.00 mm
    4 mm
    Silica
    Each
    Each - $503.50
    $503.50
    Each
       $503.50
     : 18206
    New Product
    New Product
  • Guard filter, for 4.6 mm ID columns (Super-ODS, -Octyl, -Phenyl)
    C18 (ODS or Octadecyl)
    Acier inoxydable
    2 µm
    4.00 mm
    4 mm
    Silica
    Each
    Each - $762.06
    $762.06
    Each
       $762.06
     : 18207
    New Product
    New Product
Guard Cartridge
  • Désignation
    Phase
    Taille de pore
    Column Hardware
    Granulométrie
    Ø int.
    Longueur
    Packing Material
  • TSKgel Octadecyl-4PW Guard cartridge, for 4.6 mm ID Columns
    Acier inoxydable
    7 µm
    3.20 mm
    15 mm
    Each
    Each - $745.72
    $745.72
    Each
       $745.72
     : 19008