Two-Position Snap Caps, Simport Scientific
Two-Position Snap Caps, Simport Scientific
Supplier:   Simport Scientific
Two-Position Snap Caps, Simport Scientific
Supplier:  Simport Scientific
Certificates
Two-Position Snap Caps, Simport Scientific
Two-Position Snap Caps, Simport Scientific
4 Options Available Below
Product Details & Documents
Dual position caps are suitable for use in both aerobic and anaerobic procedures.
  • Cap for aerobic and anaerobic procedures
  • For use with 12 and 17 mm tubes
  • Polystyrene and polypropylene options available
Caps offer two possibilities. The closed but unsealed position where samples are maintained aerobic for microbiological procedures; and the fully sealed position where the cap is pushed tight to seal the tube for anaerobic use or for storage, transfer and centrifuge applications. Not autoclavable.
Product Family Options
Informations sur le produit
  • Désignation
    For Tubes
    Color
    Availability
    Price
  • Two-Position Snap Caps
    12 mm Polystyrene Tubes
    Natural
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $134.64
    $134.64
    Case of 1,000
       $134.64
    Supplier Number: T401-3DSPE
  • Two-Position Snap Caps
    12 mm Polypropylene Tubes
    Natural
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $103.22
    $103.22
    Case of 1,000
       $103.22
    Supplier Number: T401-3DSPP
  • Two-Position Snap Caps
    17 mm Polystyrene Tubes
    Natural
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $147.42
    $147.42
    Case of 1,000
       $147.42
    Supplier Number: T401-10DSPE
  • Two-Position Snap Caps
    17 mm Polypropylene Tubes
    Natural
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $131.54
    $131.54
    Case of 1,000
       $131.54
    Supplier Number: T401-10DSPP