Screw Caps, PerkinElmer
Catalog # CA97019-456
Supplier: PerkinElmer
Specifications
- Color:Black
- Description:Screw Cap, Open Top, for Standard Opening 8 mm Vials
- Material:PP
- Cat. no.:CA97019-456
- Supplier No.:N9303441
Specifications
About this item
Screw caps are available in a variety of different plastics and are compatible with screwthread vials.
- Screw top design
- Inert
- Lined caps provide a tight seal
- Can be color-coded for quick identification
Screw caps provide a tight seal for protecting against volatile chemicals in autosampler applications.