Nebulizer Cleaning Wires, PerkinElmer
CA97013-702
- Description:Nebulizer Cleaning Wires
- Cat. no.:CA97013-702
- Supplier No.:03030135
Made of soft alloy copper, these cleaning wires are specially deburred to prevent scratching of the orifices and internal components of the nebulizers while cleaning.
- Soft copper construction
- Deburred
- Provided in convenient packs of multiples
Use only PerkinElmer cleaning wires to clean nebulizers.