Polysoft Face Shields, OP-D-OP
 :   OP-D-OP
Polysoft Face Shields, OP-D-OP
 :  OP-D-OP
Polysoft Face Shields, OP-D-OP
Face shield is designed for facial protection in medical facilities and laboratories.
  • Unbreakable polycarbonate shield can be replaced
  • Cinch cord can be adjusted for a secure fit
  • Crystal clear optics
  • Polysoft band for comfort
Fits over glasses and replaces goggles and masks.
Product Family Options
  • Désignation
    Color
  • Polysoft Face Shield
    Blue
    Each
    Each - $145.61
    $145.61
    Each
       $145.61
     : 206PVRB
  • Polysoft Face Shield
    Yellow
    Each
    Each - $198.92
    $198.92
    Each
       $198.92
     : 206PVNY
  • Polysoft Face Shield
    Magenta
    Each
    Each - $198.92
    $198.92
    Each
       $198.92
     : 206PVMG