Rigid-Lock QUICKBERM®, Justrite® | Avantor
Rigid-Lock QUICKBERM®, Justrite®
Supplier:   Justrite
Rigid-Lock QUICKBERM®, Justrite®
Supplier:  Justrite
Certificates
Rigid-Lock QUICKBERM®, Justrite®
Rigid-Lock QUICKBERM®, Justrite®
3 Options Available Below
Product Details & Documents
Ergonomic design provides 100% inside berm accessibility with an unobstructed side-to-side floor space for vehicle tires, foot traffic, and cleaning.
  • Single-piece construction - patented Rigid-Lock wall-support system integral to berm provides unobstructed work space
  • Quick and easy setup - no assembly required
  • Durable EnGuard™V, modified PVC coated fabric withstands long-term UV exposure
Exceptionally strong structural integrity includes a reinforced CriticalCorner™ design featuring wraparound diagonal radio frequency welds to prevent billowing, unwrapping, or sagging of the corners during full containment for fluid-tight reliability. Sidewalls lower quickly for easy drive-in and out. Portable design folds down quickly for storage and transport.
The built-in Rigid-Lock wall supports maximize usable space inside the berm and greatly reduce any tripping hazard on the berm perimeter. The device only folds down one way, inward towards the inside of the berm. The one-piece design folds down flat and remains integral to the sidewall. Supports resist extreme temperatures (-50 to 160ºF). When in the down position, they can withstand vehicle weights in excess of 11,000 lbs (5,000 kg) at each tire
Product Family Options
Informations sur le produit
  • Matière
    Folded Size
    Interior Dimensions
    Exterior Dimensions
    Sump Capacity
    Availability
    Price
  • EnGuard™V, modified PVC coated fabric
    66L x 66W x 30.5H cm (26 x 26 x 12")
    243.8W x 243.8D x 30.5H cm (96 x 96 x 12")
    264.2L x 264.2W x 30.5H cm (104 x 104 x 12")
    1798 L (475 gal.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (475US Gal)
    Each (475US Gal) - $2,483.95
    $2,483.95
    Prix promotionnel pour les nouveaux clients
    Each (475US Gal)
       $2,483.95
    Supplier Number: 28518
  • EnGuard™V, modified PVC coated fabric
    71.1L x 71.1W x 30.5H cm (28 x 28 x 12")
    365.8W x 365.8D x 30.5H cm (144 x 144 x 12")
    386.1L x 386.1W x 30.5H cm (152 x 152 x 12")
    4069 L (1075 gal.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (1,075US Gal)
    Each (1,075US Gal) - $3,001.85
    $3,001.85
    Prix promotionnel pour les nouveaux clients
    Each (1,075US Gal)
       $3,001.85
    Supplier Number: 28522
  • EnGuard™V, modified PVC coated fabric
    91.4L x 91.4W x 40.6H cm (36 x 36 x 16")
    487.7W x 487.7D x 30.5H cm (192 x 192 x 12")
    508L x 508W x 30.5H cm (200 x 200 x 12")
    7230 L (1910 gal.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (1,910US Gal)
    Each (1,910US Gal) - $5,918.61
    $5,918.61
    Each (1,910US Gal)
       $5,918.61
    Supplier Number: 28528