HLT Titertek 200 Specialty Tips, Molecular BioProducts
Catalog # 72830-676
Supplier: Thermo Scientific
Specifications
- Volume:200 µl
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:ISO14001 (Certificate Registration No. EMS 752603)
- Tip Color:Clear
- Packed:Bulk, 1000 Tips/bag
- Cat. no.:72830-676
- Tip Style:HLT Titertek 200
- Supplier No.:3850-11
Specifications
About this item
Titertek style pipettors 20-200 ul, conatins 1000 tips in each bag.
- Clear
- Non-sterile
- Nucleic acid and nuclease free
- Non pyrogenic