About this item
Lysing Matrix C is used primarily for lysis of yeast and fungi and can also be used for molds, spores and filamentous algae.
- Lysis of yeast and fungi
- Used for molds, spores and filamentous algae
Lysing Matrix C is used primarily for lysis of yeast and fungi and can also be used for molds, spores and filamentous algae
Each impact-resistant 2 mL tube contains 1 mm silica spheres. Lysing Matrix C tubes have red caps and are found in the FastRNA Pro Red Kit and FastProtein Red Matrix. Lysing Matrix C is used primarily for lysis of yeast and fungi and can also be used for molds, spores and filamentous algae.
This grinding matrix is chemically inert and will not bind nucleic acids.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA76046-070
- CA76046-068
- 75784-624
- 75784-622
- 75784-620
- Tube compatibility:
- 2 ml
- 2 ml
- 2 ml
- 2 mL
- 2 ml
- Specific gravity:
- 2.55
- 2.55
- 2.55
- 2.55
- 2.55
- Sample volume:
- up to 200 mg per prep
- up to 200 mg per prep
- up to 200 mg per prep
- up to 200 mg per prep
- up to 200 mg per prep
- Storage and handling:
- Store at Room Temperature (15...30 °C)
- Store at Room Temperature (15...30 °C)
- Store at Room Temperature (15...30 °C)
- Store at Room Temperature (15-30 °C
- Store at Room Temperature (15...30 °C)
- Sample type:
- Yeast; Fungi; Spores; Algae
- Yeast; Fungi; Spores; Algae
- Yeast; Fungi; Spores; Algae
- Yeast; Fungi; Spores; Algae
- Yeast; Fungi; Spores; Algae
- Product type:
- Lab Essentials
- Lab Essentials
- Lab Essentials
- Lab Essentials
- Lab Essentials
- Molecule enriched:
- DNA; RNA; Proteins; Other small molecules
- DNA; RNA; Proteins; Other small molecules
- DNA; RNA; Proteins; Other small molecules
- DNA; RNA; Proteins; Other small molecules
- DNA; RNA; Proteins; Other small molecules
- Matrix type:
- C
- C
- C
- C
- C
- Materials required (not supplied):
- FastPrep® Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
- FastPrep® Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
- FastPrep® Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
- FastPrep® Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
- FastPrep® Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
- Material of construction:
- Matrix C, contains 1 mm silica spheres
- Matrix C, contains 1 mm silica spheres
- Matrix C, contains 1 mm silica spheres
- Matrix C, contains 1 mm silica spheres
- Matrix C, contains 1 mm silica spheres
- Key applications:
- Extractions of nucleic acids, proteins, metabolites and other small molecules from yeast, fungi and algal samples
- Extractions of nucleic acids, proteins, metabolites and other small molecules from yeast, fungi and algal samples
- Extractions of nucleic acids, proteins, metabolites and other small molecules from yeast, fungi and algal samples
- Extractions of nucleic acids, proteins, metabolites and other small molecules from yeast, fungi and algal samples
- Extractions of nucleic acids, proteins, metabolites and other small molecules from yeast, fungi and algal samples
- Instrument Compatibility:
- FastPrep-24; FastPrep 120; FastPrep-96; SuperFastPrep-2
- FastPrep-24; FastPrep 120; FastPrep-96; SuperFastPrep-2
- FastPrep-24; FastPrep 120; FastPrep-96; SuperFastPrep-2
- FastPrep-24; FastPrep 120; FastPrep-96; SuperFastPrep-2
- FastPrep-24; FastPrep 120; FastPrep-96; SuperFastPrep-2
- Adapter compatibility:
- QuickPrep (24×2 ml), HiPrep (48×2 ml), CoolPrep (24×2 ml), QuickFlex (96×2 ml), ConeFlex Legacy (allows all FP24 adapters on FP96)
- QuickPrep (24×2 ml), HiPrep (48×2 ml), CoolPrep (24×2 ml), QuickFlex (96×2 ml), ConeFlex Legacy (allows all FP24 adapters on FP96)
- QuickPrep (24×2 ml), HiPrep (48×2 ml), CoolPrep (24×2 ml), QuickFlex (96×2 ml), ConeFlex Legacy (allows all FP24 adapters on FP96)
- QuickPrep (24x2 mL); HiPrep (48x2 mL); CoolPrep (24x2 mL); QuickFlex (96x2 mL); ConeFlex Legacy (allows all FP24 adapters on FP96)
- QuickPrep (24×2 ml), HiPrep (48×2 ml), CoolPrep (24×2 ml), QuickFlex (96×2 ml), ConeFlex Legacy (allows all FP24 adapters on FP96)
- Application areas:
- Sample Preparation; Sample Lysis; Nucleic Acid Isolation & Purification; Molecular Biology; Gene Expression; Microbiological Analysis; Gene Function; Gene Regulation
- Sample Preparation; Sample Lysis; Nucleic Acid Isolation & Purification; Molecular Biology; Gene Expression; Microbiological Analysis; Gene Function; Gene Regulation
- Sample Preparation; Sample Lysis; Nucleic Acid Isolation & Purification; Molecular Biology; Gene Expression; Microbiological Analysis; Gene Function; Gene Regulation
- Sample Preparation; Sample Lysis; Nucleic Acid Isolation & Purification; Molecular Biology; Gene Expression; Microbiological Analysis; Gene Function; Gene Regulation
- Sample Preparation; Sample Lysis; Nucleic Acid Isolation & Purification; Molecular Biology; Gene Expression; Microbiological Analysis; Gene Function; Gene Regulation