About this item
Lysing matrix D is for rapid and reliable biological sample lysis from a wide variety of starting materials.
- Impact resistant tube containing 1.4mm ceramic spheres
- Primarily used for the lysis of softer tissues
- Will also lyse plant tissue, cultured cells and whole insects
- Grinding matrix is chemically inert and will not bind nucleic acids
Lysing Matrices are critical components of the FastPrep™ Sample Preparation system. MP Biomedicals recieves the matrix particles from proven vendors and ensures the unprocessed material meets high qualitative standards. Matrix particles are then prepared and dispensed under a rigorous set of proprietary conditions that offer customers a premium product that can be immediately used with confidence.Matrices are available separately for use with your own unique buffers, and are also available as components of the complete purification kits.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- IC116913100
- CA76046-072
- CA76045-736
- CA76045-734
- 75784-626
- CA76045-776
- CA76045-774
- CA76046-058
- 75784-606
- CA76046-190
- Dimensions:
- 1.2 - 1.6 mm
- 1.2 - 1.6 mm
- 1.2 - 1.6 mm
- 1.2 - 1.6 mm
- 1.2 - 1.6 mm
- 1.2 - 1.6 mm
- 1.2 - 1.6 mm
- 1.2 - 1.6 mm
- 1.2 - 1.6 mm
- 1.2 - 1.6 mm
- Tube compatibility:
- 2 mL
- 2 ml
- 4.5 ml
- 4.5 ml
- 2 ml
- 1.2 ml
- 1.2 ml
- 2 ml
- 2 ml
- 15 ml
- Specific gravity:
- 4
- 4
- 4
- 4
- 4
- 4
- 4
- 4
- 4
- 4
- Storage and handling:
- Store at Room Temperature (15 to 30° C)
- Store at Room Temperature (15...30 °C)
- Store at Room Temperature (15...30 °C)
- Store at Room Temperature (15...30 °C)
- Store at Room Temperature (15...30 °C)
- Store at Room Temperature (15...30 °C)
- Store at Room Temperature (15...30 °C)
- Store at Room Temperature (15...30 °C)
- Store at Room Temperature (15...30 °C)
- Store at Room Temperature (15...30 °C)
- Sample type:
- Plants; Animal Tissues; Cultured Cells
- Plants; Animal Tissues; Cultured Cells
- Plants; Animal Tissues; Cultured Cells
- Plants; Animal Tissues; Cultured Cells
- Plants; Animal Tissues; Cultured Cells
- Plants; Animal Tissues; Cultured Cells
- Plants; Animal Tissues; Cultured Cells
- Plants; Animal Tissues; Cultured Cells
- Plants; Animal Tissues; Cultured Cells
- Plants; Animal Tissues; Cultured Cells
- Product type:
- Lab Essentials
- Lab Essentials
- Lab Essentials
- Lab Essentials
- Lab Essentials
- Lab Essentials
- Lab Essentials
- Lab Essentials
- Lab Essentials
- Lab Essentials
- Molecule enriched:
- DNA; RNA; Proteins; Other small molecules
- DNA; RNA; Proteins; Other small molecules
- DNA; RNA; Proteins; Other small molecules
- DNA; RNA; Proteins; Other small molecules
- DNA; RNA; Proteins; Other small molecules
- DNA; RNA; Proteins; Other small molecules
- DNA; RNA; Proteins; Other small molecules
- DNA; RNA; Proteins; Other small molecules
- DNA; RNA; Proteins; Other small molecules
- DNA; RNA; Proteins; Other small molecules
- Matrix type:
- D
- D
- D
- D
- D
- D
- D
- D
- D
- D
- Materials required (not supplied):
- FastPrep® Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
- FastPrep® Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
- FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
- FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
- FastPrep® Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
- FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
- FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
- FastPrep® Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
- FastPrep® Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
- FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
- Material of construction:
- Matrix D, contains 1.4 mm ceramic spheres.
- Matrix D, contains 1.4 mm ceramic spheres.
- 1.4 mm ceramic spheres
- 1.4 mm ceramic spheres
- Matrix D, contains 1.4 mm ceramic spheres.
- 1.4 mm ceramic spheres
- 1.4 mm ceramic spheres
- Matrix D, contains 1.4 mm ceramic spheres.
- Matrix D, contains 1.4 mm ceramic spheres.
- Matrix D, contains 1.4 mm ceramic spheres
- Key applications:
- Lysis of soft tissues
- Lysis of soft tissues
- Lysis of soft tissues
- Lysis of soft tissues
- Lysis of soft tissues
- Lysis of soft tissues
- Lysis of soft tissues
- Lysis of soft tissues
- Lysis of soft tissues
- Lysis of soft tissues
- Instrument Compatibility:
- FastPrep-24; FastPrep 120; FastPrep-96
- FastPrep-24; FastPrep 120; FastPrep-96
- FastPrep-24, FastPrep-96, FastPrep-24 5G
- FastPrep-24, FastPrep-96, FastPrep-24 5G
- FastPrep-24; FastPrep 120; FastPrep-96
- FastPrep-96
- FastPrep-96
- FastPrep-24; FastPrep 120; FastPrep-96
- FastPrep-24; FastPrep 120; FastPrep-96
- FastPrep-24; FastPrep-96
- Adapter compatibility:
- QuickPrep (24x2 mL); HiPrep (48x2 mL); CoolPrep (24x2 mL); QuickFlex (96x2 mL); ConeFlex Legacy (allows all FP24 adapters on FP96)
- QuickPrep (24×2 ml), HiPrep (48×2 ml), CoolPrep (24×2 ml), QuickFlex (96×2 ml), ConeFlex Legacy (allows all FP24 adapters on FP96)
- TallPrep (24×4.5 ml), TallFlex (48×4.5 ml), ConeFlex Legacy (allows all FP24 adapters on FP96)
- TallPrep (24×4.5 ml), TallFlex (48×4.5 ml), ConeFlex Legacy (allows all FP24 adapters on FP96)
- QuickPrep (24×2 ml), HiPrep (48×2 ml), CoolPrep (24×2 ml), QuickFlex (96×2 ml), ConeFlex Legacy (allows all FP24 adapters on FP96)
- 96 Well Adapter
- 96 Well Adapter
- QuickPrep (24×2 ml), HiPrep (48×2 ml), CoolPrep (24×2 ml), QuickFlex (96×2 ml), ConeFlex Legacy (allows all FP24 adapters on FP96)
- QuickPrep (24×2 ml), HiPrep (48×2 ml), CoolPrep (24×2 ml), QuickFlex (96×2 ml), ConeFlex Legacy (allows all FP24 adapters on FP96)
- TeenPrep (12×15 ml), CoolTeenPrep (6×15 ml), TeenFlex (24×15 ml), ConeFlex Legacy (allows all FP24 adapters on FP96)
- Application areas:
- Nucleic Acid Isolation & Purification
- Nucleic Acid Isolation & Purification
- Nucleic Acid Isolation & Purification
- Nucleic Acid Isolation & Purification
- Nucleic Acid Isolation & Purification
- Nucleic Acid Isolation & Purification
- Nucleic Acid Isolation & Purification
- Nucleic Acid Isolation & Purification
- Nucleic Acid Isolation & Purification
- Nucleic Acid Isolation & Purification