Well Plates, Restek
: Restek
Polypropylene plates with round-bottom wells reduce liquid retention; conical bottom provides optimal recovery of reagents.
- Round well shape is ideal for applications that require vortexing
- Ideal for sample collection, storage, sampling, and combinatorial chemistry and library applications
- Fit most autosampler compartments
Nunc® shared wall technology allows increased well volume for optimum storage capacity and improved mixing.
: All microplates manufactured by Nunc® meet the recommendation of American National Standards Institute (ANSI) (ANSI/SBS 1-2004).
Polypropylene plates with round-bottom wells reduce liquid retention; conical bottom provides optimal recovery of reagents.
- Round well shape is ideal for applications that require vortexing
- Ideal for sample collection, storage, sampling, and combinatorial chemistry and library applications
- Fit most autosampler compartments
Nunc® shared wall technology allows increased well volume for optimum storage capacity and improved mixing.
: All microplates manufactured by Nunc® meet the recommendation of American National Standards Institute (ANSI) (ANSI/SBS 1-2004).
Product Family Options
- DésignationWell Bottom ShapeWell Shape
- 0.45 mL 96-Well PlatesConicalRoundPack of 120$1,293.93: 26496
- : 26492
- : 26494
- : 26497
- : 26493
- : 26495