Extended Capillary Column Nuts for Agilent 5890/6850/6890/7890 GCs, Restek
Fournisseur: Restek
Certificats
À propos de cet article
Visible below inlet insulating cup for easy, breakage-free column installation without removing cup.
- Finger-tight design to allow wrench-free use
- Available for standard (1/16") and compact (Agilent-style) ferrules
- Made from high-quality stainless steel
- Meets or exceeds original manufacturer's performance
Frequently Bought Together