Replacement Septa, PTFE-Lined Silicone for VOA Sampling Vials, 60 mL Sample Collection Vials, and 250 ml Sample Collection Bottles, Restek
10854-242
: Restek
- Ø:24 mm
- Description:Replacement septa
- Thickness:0.125"
- Cat. no.:10854-242
- Supplier No.:24694
These replacement septa are extremely economical accessories for use with various sample collection vials and bottles.
- Compatible with many vials
- Economical
- PTFE-lined silicone construction
PTFE-lined silicone replacement septa are used with VOA sampling vials, 60 ml sample collection vials, and 250 ml sample collection bottles.