Replacement Microseal Nut for Merlin Microseal Septa for Agilent GCs, Restek
Catalog # RK22809
Supplier: Restek
Specifications
- Description:Replacement Microseal Nut
- Environmentally Preferable:
- Cat. no.:RK22809
- Supplier No.:22809
Specifications
About this item
Replacement microseal nuts are available for use with "septum-less" Merlin microseal systems, providing an extra layer of sealing.
- Effective seal
- Durable and non-contaminating
These microseal nuts are designed to maintain a leak-free seal without the use of a septum, so septum fragments will not be formed and deposited in the inlet liner.