Standard Ferrules for PerkinElmer GCs, Fit 0.25/0.32 mm int.Ø MXT® Columns, Restek
Fournisseur: Restek
Certificats
À propos de cet article
Standard ferrules fit ID MXT® columns with internal diameters of 0.25 mm or 0.32 mm
- VG2, 60% Vespel®/40% graphite blend offers the best combination of sealing and ease of workability
- VG1, 85% Vespel®/15% graphite blend offers equivalent composition to Agilent Vespel®/graphite ferrules
- Seal with minimal torque, reusable, and preferred for vacuum and high-pressure uses
- Stable to 400°C
- Recommended for mass spec transfer lines
For use with PerkinElmer GCs.
Frequently Bought Together