SPI with Buffer for Bruker/Varian GCs equipped with SPI inlets, Restek
Catalog # RK20851
Supplier: Restek
Specifications
- Ext.Ø:4.6 mm
- Volume:0.141 ml
- Description:SPI with Buffer
- Material:Borosilicate glass
- Cat. no.:RK20851
- For Use With:Bruker/Varian
- I.D.×L:2.4×54 mm
- Treatment:Intermediate polarity (IP)
- Supplier No.:20851
Specifications
About this item
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
- Borosilicate glass fabrication
- Consistent quality with exceptional batch-to-batch uniformity
- Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.